missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CASP7 / caspase 7 / MCH3 Protein

A DNA sequence encoding the Homo sapiens (Human) CASP7 / caspase 7 / MCH3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8834-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016731

  • 2090.00 EUR / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001218.1
CASP7 / caspase 7 / MCH3 Protein
Research Use Only
CASP7
Recombinant Protein
E. coli
ANPRYKIPVE ADFLFAYSTV PGYYSWRSPG RGSWFVQALC SILEEHGKDL EIMQILTRVN DRVARHFESQ SDDPHFHEKK QIPCVVSMLT KELYFSQ
Greater than 90% as determined by SDS-PAGE.
840
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers