missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase II subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase II subunit beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8381-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Casein kinase II subunit beta Protein | |
Research Use Only | |
CSNK2B | |
Recombinant Protein | |
E. coli | |
SSSEEVSWIS WFCGLRGNEF FCEVDEDYIQ DKFNLTGLNE QVPHYRQALD MILDLEPDEE LEDNPNQSDL IEQAAEMLYG LIHARYILTN RGIAQMLEKY QQGDFGYCPR VYCENQPMLP IGLSDIPGEA MVKLYCPKCM DVYTPKSSRH HHTDGAYFGT GFPHMLFMVH PEYRPKRPAN QFVPRLYGFK IHPMAYQLQL QAASNFKSPV KTIR | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |