missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein

A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8358-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034211

  • 2090.00 EUR / 1mg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001276841.1
Casein kinase I isoform epsilon Protein
Research Use Only
CSNK1E
Recombinant Protein
E. coli
SGSFGDIYLG ANIASGEEVA IKLECVKTKH PQLHIESKFY KMMQGGVGIP SIKWCGAEGD YNVMVMELLG PSLEDLFNFC SRKFSLKTVL LLADQMISRI EYIHSKNFIH RDVKPDNFLM GLGKKGNLVY IIDFGLAKKY RDARTHQHIP YRENKNLTGT ARYASINTHL GIEQSRRDDL ESLGYVLMYF NLGSLPWQGL KAATKRQKYE RISEK
Greater than 90% as determined by SDS-PAGE.
102800317
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers