missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Carbonyl reductase [NADPH] 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonyl reductase [NADPH] 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8781-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273718.1 | |
Carbonyl reductase [NADPH] 1 Protein | |
Research Use Only | |
CBR1 | |
Recombinant Protein | |
Yeast | |
SSGIHVALVT GGNKGIGLAI VRDLCRLFSG DVVLTARDVT RGQAAVQQLQ AEGLSPRFHQ LDIDDLQSIR ALRDFLRKEY GGLDVLVNNA GIAFKVADPT PFHIQAEVTM KTNFFGTRDV CTELLPLIKP QGRVVNVSSI MSVRALKSCS PELQQKFRSE TITEEELVGL MNKFVEDTKK GVHQKEGWPS SAYGVTKIGV TVLSRIHARK LSEQRKGDKI LLNACCPGWV RTDMAGPKAT KSPEEGAETP VYLALLPPDA EGPHGQFVSE KRVEQW | |
Greater than 90% as determined by SDS-PAGE. |
873 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |