missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cancer / testis antigen 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cancer/testis antigen 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9084-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001318.1 | |
Cancer / testis antigen 1 Protein | |
Research Use Only | |
CTAG1A | |
Recombinant Protein | |
Yeast | |
MQAEGRGTGG STGDADGPGG PGIPDGPGGN AGGPGEAGAT GGRGPRGAGA ARASGPGGGA PRGPHGGAAS GLNGCCRCGA RGPESRLLEF YLAMPFATPM EAELARRSLA QDAPPLPVPG VLLKEFTVSG NILTIRLTAA DHRQLQLSIS SCLQQLSLLM WITQCFLPVF LAQPPSGQRR | |
Greater than 90% as determined by SDS-PAGE. |
1485 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |