missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calponin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Calponin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8036-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001290428.1 | |
Calponin-2 Protein | |
Research Use Only | |
CNN2 | |
Recombinant Protein | |
E. coli | |
SSTQFNKGPS YGLSAEVKNR LLSKYDPQKE AELRTWIEGL TGLSIGPDFQ KGLKDGTILC TLMNKLQPGS VPKINRSMQN WHQLENLSNF IKAMVSYGMN PVDLFEANDL FESGNMTQVQ VSLLALAGKA KTKGLQSGVD IGVKYSEKQE RNFDDATMKA GQCVIGLQMG TNKCASQSGM TAYGTRRHLY DPKNHILPPM DHSTISLQMG TNKCASQVGM TAPGTRRHIY DTKLGTDKCD NSSMSLQMGY TQGANQSGQV FGLGRQIYDP KYCPQGTVAD GAPSGTGDCP DPGEVPEYPP YYQEEAGY | |
Greater than 90% as determined by SDS-PAGE. |
1265 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |