missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bone morphogenetic protein 1

A DNA sequence encoding the Homo sapiens (Human) Bone morphogenetic protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8701-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016031

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001709.1
Bone morphogenetic protein 1
Research Use Only
BMP6
Recombinant Protein
E. coli
QQSRNRSTQS QDVARVSSAS DYNSSELKTA CRKHELYVSF QDLGWQDWII APKGYAANYC DGECSFPLNA HMNATNHAIV QTLVHLMNPE YVPKPCCAPT KLNAISVLYF DDNSNVILKK YRNMVVRACG CH
Greater than 90% as determined by SDS-PAGE.
654
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers