missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bone morphogenetic protein 1
A DNA sequence encoding the Homo sapiens (Human) Bone morphogenetic protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8701-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001709.1 | |
Bone morphogenetic protein 1 | |
Research Use Only | |
BMP6 | |
Recombinant Protein | |
E. coli | |
QQSRNRSTQS QDVARVSSAS DYNSSELKTA CRKHELYVSF QDLGWQDWII APKGYAANYC DGECSFPLNA HMNATNHAIV QTLVHLMNPE YVPKPCCAPT KLNAISVLYF DDNSNVILKK YRNMVVRACG CH | |
Greater than 90% as determined by SDS-PAGE. |
654 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |