missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BOLA1 Protein
A DNA sequence encoding the Homo sapiens (Human) BOLA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9011-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307954.1 | |
BOLA1 Protein | |
Research Use Only | |
BOLA1 | |
Recombinant Protein | |
Yeast | |
MLSGRLVLGL VSMAGRVCLC QGSAGSGAIG PVEAAIRTKL EEALSPEVLE LRNESGGHAV PPGSETHFRV AVVSSRFEGL SPLQRHRLVH AALAEELGGP VHALAIQART PAQWRENSQL DTSPPCLGGN KKTLGTP | |
Greater than 90% as determined by SDS-PAGE. |
51027 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |