missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLyS / TNFSF13B / BAFF Protein
A DNA sequence encoding the Homo sapiens (Human) BLyS / TNFSF13B / BAFF, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001139117.1 |
---|---|
Gene ID (Entrez) | 10673 |
Name | BLyS / TNFSF13B / BAFF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033861
|
enQuireBio™
QP8298-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053861
|
enQuireBio™
QP8298-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043861
|
enQuireBio™
QP8298-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063861
|
enQuireBio™
QP8298-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073861
|
enQuireBio™
QP8298-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083861
|
enQuireBio™
QP8298-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001139117.1 | |
BLyS / TNFSF13B / BAFF Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10673 | |
Research Use Only | |
TNFSF13B | |
Recombinant Protein | |
E. coli | |
QVAALQGDLA SLRAELQGHH AEKLPAGAGA PKAGLEEAPA VTAGLKIFEP PAPGEGNSSQ NSRNKRAVQG PEETVTQDCL QLIADSETPT IQKGSYTFVP WLLSFKRGSA LEEKENKILV KETGYFFIYG QVLYTDKTYA MGHLIQRKKV HVFGDELSLV TLFRCIQNMP ETLPNNSCYS AGIAKLEEGD ELQLAIPREN AQISLDGDVT FFGALKLL | |
Greater than 90% as determined by SDS-PAGE. |