Learn More
enQuireBio™ Recombinant Human BLID Protein
A DNA sequence encoding the Homo sapiens (Human) BLID, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7821-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001786.2 | |
BLID Protein | |
Research Use Only | |
BLID | |
Recombinant Protein | |
E. coli | |
MVTLLPIEGQ EIHFFEILES ECVLYTGWIE RASGSSIYPE AKARLPLEAL LGSNKEPMLP KETVLSLKRY NLGSSAMKRN VPGHVLQRPS YLTRIQVTLL CNSSAEAL | |
Greater than 80% as determined by SDS-PAGE. |
414899 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |