missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAG family molecular chaperone regulator 5 Protein
A DNA sequence encoding the Homo sapiens (Human) BAG family molecular chaperone regulator 5, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001015048.1 |
---|---|
Gene ID (Entrez) | 9529 |
Name | BAG family molecular chaperone regulator 5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023451
|
enQuireBio™
QP8234-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043451
|
enQuireBio™
QP8234-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033451
|
enQuireBio™
QP8234-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053451
|
enQuireBio™
QP8234-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063451
|
enQuireBio™
QP8234-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073451
|
enQuireBio™
QP8234-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001015048.1 | |
BAG family molecular chaperone regulator 5 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9529 | |
Research Use Only | |
BAG5 | |
Recombinant Protein | |
E. coli | |
MDMGNQHPSI SRLQEIQKEV KSVEQQVIGF SGLSDDKNYK KLERILTKQL FEIDSVDTEG KGDIQQARKR AAQETERLLK ELEQNANHPH RIEIQNIFEE AQSLVREKIV PFYNGGNCVT DEFEEGIQDI ILRLTHVKTG GKISLRKARY HTLTKICAVQ EIIEDCMKKQ PSLPLSEDAH PSVAKINFVM CEVNKARGVL IALLMGVNNN ETCRHLSCVL SGLIADLDAL DVCGRTEIRN YRREVVEDIN KLLKYLDLEE EADTTKAFDL RQNHSILKIE KVLKRMREIK NELLQAQNPS ELYLSSKTEL QGLIGQLDEV SLEKNPCIRE ARRRAVIEVQ TLITYIDLKE ALEKRKLFAC EEHPSHKAVW NVLGNLSEIQ GEVLSFDGNR TDKNYIRLEE LLTKQLLALD AVDPQGEEKC KAARKQAVRL AQNILSYLDL KSDEWEY | |
Greater than 90% as determined by SDS-PAGE. |