missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP-dependent RNA helicase DDX19A Protein
A DNA sequence encoding the Homo sapiens (Human) ATP-dependent RNA helicase DDX19A, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001307451.1 |
---|---|
Gene ID (Entrez) | 55308 |
Name | ATP-dependent RNA helicase DDX19A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023391
|
enQuireBio™
QP8225-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043391
|
enQuireBio™
QP8225-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033391
|
enQuireBio™
QP8225-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053391
|
enQuireBio™
QP8225-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063391
|
enQuireBio™
QP8225-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073391
|
enQuireBio™
QP8225-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001307451.1 | |
ATP-dependent RNA helicase DDX19A Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
55308 | |
Research Use Only | |
DDX19A | |
Recombinant Protein | |
E. coli | |
MATDSWALAV DEQEAAVKSM TNLQIKEEKV KADTNGIIKT STTAEKTDEE EKEDRAAQSL LNKLIRSNLV DNTNQVEVLQ RDPNSPLYSV KSFEELRLKP QLLQGVYAMG FNRPSKIQEN ALPMMLAEPP QNLIAQSQSG TGKTAAFVLA MLSRVEPSDR YPQCLCLSPT YELALQTGKV IEQMGKFYPE LKLAYAVRGN KLERGQKISE QIVIGTPGTV LDWCSKLKFI DPKKIKVFVL DEADVMIATQ GHQDQSIRIQ RMLPRNCQML LFSATFEDSV WKFAQKVVPD PNVIKLKREE ETLDTIKQYY VLCSSRDEKF QALCNLYGAI TIAQAMIFCH TRKTASWLAA ELSKEGHQVA LLSGEMMVEQ RAAVIERFRE GKEKVLVTTN VCARGIDVEQ VSVVINFDLP VDKDGNPDNE TYLHRIGRTG RFGKRGLAVN MVDSKHSMNI LNRIQEHFNK KIERLDTDDL DEIEKIAN | |
Greater than 90% as determined by SDS-PAGE. |