missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Aryl-hydrocarbon-interacting protein-like 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Aryl-hydrocarbon-interacting protein-like 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8180-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033111

  • 675.00 EUR / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001028226.1
Aryl-hydrocarbon-interacting protein-like 1 Protein
Research Use Only
AIPL1
Recombinant Protein
E. coli
MDAALLLNVE GVKKTILHGG TGELPNFITG SRVIFHFRTM KCDEERTVID DSRQVGQPMH IIIGNMFKLE VWEILLTSMR VHEVAEFWCD TIHTGVYPIL SRSLRQMAQG KDPTEWHVHT CGLANMFAYH TLGYEDLDEL QKEPQPLVFV IELLQVDAPS DYQRETWNLS NHEKMKAVPV LHGEGNRLFK LGRYEEASSK YQEAIICLRN LQTKEKPWEV QWLKLEKMIN TLILNYCQCL LKKEEYYEVL EHTSDILRHH PGIVKAYYVR ARAHAEVWNE AEAKADLQKV LELEPSMQKA VRRELRLLEN RMAEKQEEER LRCRNMLSQG ATQPPAEPPT EPPAQSSTEP PAEPPTAPSA ELSAGPPAEP ATEPPPSPGH SLQH
Greater than 80% as determined by SDS-PAGE.
23746
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers