missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ARNT Protein

A DNA sequence encoding the Homo sapiens (Human) ARNT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8996-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047431

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001184254.1
ARNT Protein
Research Use Only
ARNT
Recombinant Protein
Yeast
MAATTANPEM TSDVPSLGPA IASGNSGPGI QGGGAIVQRA IKRRPGLDFD DDGEGNSKFL RCDDDQMSND KERFARSDDE QSSADKERLA RENHSEIERR RRNKMTAYIT ELSDMVPTCS ALARKPDKLT ILRMAVSHMK SLRGTGNTST DGSYKPSFLT DQELKHLILE AADGFLFIVS CETGRVVYVS DSVTPVLNQP QSEWFGSTLY DQVHPDDVDK LREQLSTSEN ALTGRILDLK TGTVKKEGQQ SSMRMCMGSR RSFICRMRCG SSSVDPVSVN RLSFVRNRCR NGLGSVKDGE PHFVVVHCTG YIKAWPPAGV SLPDDDPEAG QGSKFCLVAI GRLQVTSSPN CTDMSNVCQP TEFISRHNIE GIFTFVDHRC VATVGYQPQE LLGKNIVEFC HPEDQQLLRD SFQQVVKLKG QVLSVMFRFR SKNQEWLWMR TSSFTFQNPY SDEIEYIICT NTNVKNSSQE PRPT
Greater than 90% as determined by SDS-PAGE.
405
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers