missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARF5 / ADP-ribosylation factor 5 Protein
A DNA sequence encoding the Homo sapiens (Human) ARF5 / ADP-ribosylation factor 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8337-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001653.1 | |
ARF5 / ADP-ribosylation factor 5 Protein | |
Research Use Only | |
ARF5 | |
Recombinant Protein | |
E. coli | |
GLTVSALFSR IFGKKQMRIL MVGLDAAGKT TILYKLKLGE IVTTIPTIGF NVETVEYKNI CFTVWDVGGQ DKIRPLWRHY FQNTQGLIFV VDSNDRERVQ ESADELQKML QEDELRDAVL LVFANKQDMP NAMPVSELTD KLGLQHLRSR TWYVQATCAT QGTGLYDGLD WLSHELSKR | |
Greater than 90% as determined by SDS-PAGE. |
381 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |