missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANXA5 / Annexin 5 / Annexin A5 Protein

A DNA sequence encoding the Homo sapiens (Human) ANXA5 / Annexin 5 / Annexin A5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8982-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017381

  • 2325.00 EUR / 500µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001145.1
ANXA5 / Annexin 5 / Annexin A5 Protein
Research Use Only
ANXA5
Recombinant Protein
Yeast
AQVLRGTVTD FPGFDERADA ETLRKAMKGL GTDEESILTL LTSRSNAQRQ EISAAFKTLF GRDLLDDLKS ELTGKFEKLI VALMKPSRLY DAYELKHALK GAGTNEKVLT EIIASRTPEE LRAIKQVYEE EYGSSLEDDV VGDTSGYYQR MLVVLLQANR DPDAGIDEAQ VEQDAQALFQ AGELKWGTDE EKFITIFGTR SVSHLRKVFD KYMTISGFQI EETIDRETSG NLEQLLLAVV KSIRSIPAYL AETLYYAMKG AGTDDHTLIR VMVSRSEIDL FNIRKEFRKN FATSLYSMIK GDTSGDYKKA LLLLCGEDD
Greater than 89.50% as determined by SDS-PAGE.
308
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers