missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Anaphase-promoting complex subunit 10 Protein

A DNA sequence encoding the Homo sapiens (Human) Anaphase-promoting complex subunit 10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8254-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023591

  • 2090.00 EUR / 1mg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001243635.1
Anaphase-promoting complex subunit 10 Protein
Research Use Only
ANAPC10
Recombinant Protein
E. coli
TTPNKTPPGA DPKQLERTGT VREIGSQAVW SLSSCKPGFG VDQLRDDNLE TYWQSDGSQP HLVNIQFRRK TTVKTLCIYA DYKSDESYTP SKISVRVGNN FHNLQEIRQL ELVEPSGWIH VPLTDNHKKP TRTFMIQIAV LANHQNGRDT HMRQIKIYTP VEESSIGKFP RCTTIDFMMY RSIR
Greater than 80% as determined by SDS-PAGE.
10393
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales