missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alpha-crystallin B chain Protein
A DNA sequence encoding the Homo sapiens (Human) Alpha-crystallin B chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5870-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001276736.1 | |
Alpha-crystallin B chain Protein | |
Research Use Only | |
CRYAB | |
Recombinant Protein | |
E. coli | |
MDIAIHHPWI RRPFFPFHSP SRLFDQFFGE HLLESDLFPT STSLSPFYLR PPSFLRAPSW FDTGLSEMRL EKDRFSVNLD VKHFSPEELK VKVLGDVIEV HGKHEERQDE HGFISREFHR KYRIPADVDP LTITSSLSSD GVLTVNGPRK QVSGPERTIP ITREEKPAVT AAPKK | |
Greater than 90% as determined by SDS-PAGE. |
1410 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |