missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AGER / RAGE Protein
A DNA sequence encoding the Homo sapiens (Human) AGER / RAGE, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8786-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001127.1 | |
AGER / RAGE Protein | |
Research Use Only | |
AGER | |
Recombinant Protein | |
E. coli | |
AQNITARIGE PLVLKCKGAP KKPPQRLEWK LNTGRTEAWK VLSPQGGGPW DSVARVLPNG SLFLPAVGIQ DEGIFRCQAM NRNGKETKSN YRVRVYQIPG KPEIVDSASE LTAGVPNKVG TCVSEGSYPA GTLSWHLDGK PLVPNEKGVS VKEQTRRHPE TGLFTLQSEL MVTPARGGDP RPTFSCSFSP GLPRHRALRT APIQPRVWEP VPLEEVQLVV EPEGGAVAPG GTVTLTCEVP AQPSPQIHWM KDGVPLPLPP SPVLILPEIG PQDQGTYSCV ATHSSHGPQE SRAVSISIIE PGEEGPTAGS VGGSGLGTLA | |
Greater than 90% as determined by SDS-PAGE. |
177 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |