missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human aFGF / FGF1 Protein
A DNA sequence encoding the Human aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.
131.00 EUR - 675.00 EUR
Specifications
Accession Number | NP_000791.1 |
---|---|
Gene ID (Entrez) | 2246 |
Name | aFGF / FGF1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086701
|
enQuireBio™
QP883-10UG |
10 μg |
131.00 EUR
10µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006711
|
enQuireBio™
QP883-50UG |
50 μg |
341.00 EUR
50µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076701
|
enQuireBio™
QP883-100UG |
100 μg |
385.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096701
|
enQuireBio™
QP883-250UG |
250 μg |
675.00 EUR
250µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_000791.1 | |
aFGF / FGF1 Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 95% as determined by SDS-PAGE and HPLC. |
2246 | |
Research Use Only | |
FGF1 | |
The ED50 as determined by a cell proliferation assay using murine balb/c 3T3 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg in the presence of 10 ug/ml of heparin. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |