missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human aFGF / FGF1 Protein
A DNA sequence encoding the Human aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP883-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000791.1 | |
aFGF / FGF1 Protein | |
Research Use Only | |
FGF1 | |
The ED50 as determined by a cell proliferation assay using murine balb/c 3T3 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg in the presence of 10 ug/ml of heparin. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
2246 | |
50 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 95% as determined by SDS-PAGE and HPLC. |