missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Activin A / INHBA Protein
A DNA sequence encoding the Homo sapiens (Human) Activin A / INHBA Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6232-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002183.1 | |
Activin A / INHBA Protein | |
Research Use Only | |
INHBA | |
Recombinant Protein | |
E. coli | |
GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS | |
Greater than 90% as determined by SDS-PAGE. |
3624 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |