missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-related protein 2/3 complex subunit 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8041-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257368.1 | |
Actin-related protein 2 / 3 complex subunit 5 Protein | |
Research Use Only | |
ARPC5 | |
Recombinant Protein | |
E. coli | |
MSKNTVSSAR FRKVDVDEYD ENKFVDEEDG GDGQAGPDEG EVDSCLRHSI TGNMTAALQA ALKNPPINTK SQAVKDRAGS IVLKVLISFK ANDIEKAVQS LDKNGVDLLM KYIYKGFESP SDNSSAMLLQ WHEKALAAGG VGSIVRVLTA RKTV | |
Greater than 80% as determined by SDS-PAGE. |
10092 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |