missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACSF2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ACSF2, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7983-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001275897.1 | |
ACSF2, mitochondrial Protein | |
Research Use Only | |
ACSF2 | |
Recombinant Protein | |
E. coli | |
LSSREVDRMV STPIGGLSYV QGCTKKHLNS KTVGQCLETT AQRVPEREAL VVLHEDVRLT FAQLKEEVDK AASGLLSIGL CKGDRLGMWG PNSYAWVLMQ LATAQAGIIL VSVNPAYQAM ELEYVLKKVG CKALVFPKQF KTQQYYNVLK QICPEVENAQ PGALKSQRLP DLTTVISVDA PLPGTLLLDE VVAAGSTRQH LDQLQYNQQF LSCHDPINIQ FTSGTTGSPK GATLSHYNIV NNSNILGERL KLHEKTPEQL RMILPNPLYH CLGSVAGTMM CLMYGATLIL ASPIFNGKKA LEAISRERGT FLYGTPTMFV DILNQPDFSS YDISTMCGGV IAGSPAPPEL IRAIINKINM KDLVVAYGTT ENSPVTFAHF PEDTVEQKAE SVGRIMPHTE ARIMNMEAGT LAKLNTPGEL CIRGYCVMLG YWGEPQKTEE AVDQDKWYWT GDVATMNEQG FCKIVGRSKD MIIRGGENIY PAELEDFFHT HPKVQEVQVV GVKDDRMGEE ICACIRLKDG EETTVEEIKA FCKGKISHFK IPKYIVFVTN YPLTISGKIQ KFKLREQMER HLNL | |
Greater than 90% as determined by SDS-PAGE. |
80221 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |