missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Acid ceramidase Protein
A DNA sequence encoding the Homo sapiens (Human) Acid ceramidase, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001120977.1 |
---|---|
Gene ID (Entrez) | 427 |
Name | Acid ceramidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009460
|
enQuireBio™
QP7569-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16029460
|
enQuireBio™
QP7569-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019460
|
enQuireBio™
QP7569-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039460
|
enQuireBio™
QP7569-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049460
|
enQuireBio™
QP7569-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059460
|
enQuireBio™
QP7569-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001120977.1 | |
Acid ceramidase Protein | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
427 | |
Research Use Only | |
ASAH1 | |
Recombinant Protein | |
E. coli | |
QHAPPWTEDC RKSTYPPSGP TYRGAVPWYT INLDLPPYKR WHELMLDKAP VLKVIVNSLK NMINTFVPSG KIMQVVDEKL PGLLGNFPGP FEEEMKGIAA VTDIPLGEII SFNIFYELFT ICTSIVAEDK KGHLIHGRNM DFGVFLGWNI NNDTWVITEQ LKPLTVNLDF QRNNKTVFKA SSFAGYVGML TGFKPGLFSL TLNERFSING GYLGILEWIL GKKDVMWIGF LTRTVLENST SYEEAKNLLT KTKILAPAYF ILGGNQSGEG CVITRDRKES LDVYELDAKQ GRWYVVQTNY DRWKHPFFLD DRRTPAKMCL NRTSQENISF ETMYDVLSTK PVLNKLTVYT TLIDVTKGQF ETYLRDCPDP CIGW | |
Greater than 90% as determined by SDS-PAGE. |