missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AASDHPPT Protein

A DNA sequence encoding the Homo sapiens (Human) AASDHPPT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8073-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052511

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_056238.2
AASDHPPT Protein
Research Use Only
AASDHPPT
Recombinant Protein
E. coli
MVFPAKRFCL VPSMEGVRWA FSCGTWLPSR AEWLLAVRSI QPEEKERIGQ FVFARDAKAA MAGRLMIRKL VAEKLNIPWN HIRLQRTAKG KPVLAKDSSN PYPNFNFNIS HQGDYAVLAA EPELQVGIDI MKTSFPGRGS IPEFFHIMKR KFTNKEWETI RSFKDEWTQL DMFYRNWALK ESFIKAIGVG LGFELQRLEF DLSPLNLDIG QVYKETRLFL DGEEEKEWAF EESKIDEHHF VAVALRKPDG SRHQDVPSQD DSKPTQRQFT ILNFNDLMSS AVPMTPEDPS FWDCFCFTEE IPIRNGTKS
Greater than 90% as determined by SDS-PAGE.
60496
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers