missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AASDHPPT Protein
A DNA sequence encoding the Homo sapiens (Human) AASDHPPT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8073-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_056238.2 | |
AASDHPPT Protein | |
Research Use Only | |
AASDHPPT | |
Recombinant Protein | |
E. coli | |
MVFPAKRFCL VPSMEGVRWA FSCGTWLPSR AEWLLAVRSI QPEEKERIGQ FVFARDAKAA MAGRLMIRKL VAEKLNIPWN HIRLQRTAKG KPVLAKDSSN PYPNFNFNIS HQGDYAVLAA EPELQVGIDI MKTSFPGRGS IPEFFHIMKR KFTNKEWETI RSFKDEWTQL DMFYRNWALK ESFIKAIGVG LGFELQRLEF DLSPLNLDIG QVYKETRLFL DGEEEKEWAF EESKIDEHHF VAVALRKPDG SRHQDVPSQD DSKPTQRQFT ILNFNDLMSS AVPMTPEDPS FWDCFCFTEE IPIRNGTKS | |
Greater than 90% as determined by SDS-PAGE. |
60496 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |