missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L5 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8412-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024471

  • 458.00 EUR / 100µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000960.2
60S ribosomal protein L5 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6125
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPL5
Recombinant Protein
E. coli
GFVKVVKNKA YFKRYQVKFR RRREGKTDYY ARKRLVIQDK NKYNTPKYRM IVRVTNRDII CQIAYARIEG DMIVCAAYAH ELPKYGVKVG LTNYAAAYCT GLLLARRLLN RFGMDKIYEG QVEVTGDEYN VESIDGQPGA FTCYLDAGLA RTTTGNKVFG ALKGAVDGGL SIPHSTKRFP GYDSESKEFN AEVHRKHIMG QNVADYMRYL MEEDEDAYKK QFSQYIKNSV TPDMMEEMYK KAHAAIRENP VYEKKPKKEV KKKRWNRPKM SLAQKKDRVA QKKASFLRAQ ERAAES
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers