missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L35 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L35, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8470-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009140.1 | |
60S ribosomal protein L35 Protein | |
Research Use Only | |
RPL35 | |
Recombinant Protein | |
E. coli | |
AKIKARDLRG KKKEELLKQL DDLKVELSQL RVAKVTGGAA SKLSKIRVVR KSIARVLTVI NQTQKENLRK FYKGKKYKPL DLRPKKTRAM RRRLNKHEEN LKTKKQQRKE RLYPLRKYAV KA | |
Greater than 90% as determined by SDS-PAGE. |
11224 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |