missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 5'-nucleotidase Protein
A DNA sequence encoding the Homo sapiens (Human) 5'-nucleotidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8692-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001191742.1 | |
5'-nucleotidase Protein | |
Research Use Only | |
NT5 | |
Recombinant Protein | |
E. coli | |
ELTILHTNDV HSRLEQTSED SSKCVNASRC MGGVARLFTK VQQIRRAEPN VLLLDAGDQY QGTIWFTVYK GAEVAHFMNA LRYDAMALGN HEFDNGVEGL IEPLLKEAKF PILSANIKAK GPLASQISGL YLPYKVLPVG DEVVGIVGYT SKETPFLSNP GTNLVFEDEI TALQPEVDKL KTLNVNKIIA LGHSGFEMDK LIAQKVRGVD VVVGGHSNTF LYTGNPPSKE VPAGKYPFIV TSDDGRKVPV VQAYAFGKYL GYLKIEFDER GNVISSHGNP ILLNSSIPED PSIKADINKW RIKLDNYSTQ ELGKTIVYLD GSSQSCRFRE CNMGNLICDA MINNNLRHTD EMFWNHVSMC ILNGGGIRSP IDERNNGTIT WENLAAVLPF GGTFDLVQLK GSTLKKAFEH SVHRYGQSTG EFLQVGGIHV VYDLSRKPGD RVVKLDVLCT KCRVPSYDPL KMDEVYKVIL PNFLANGGDG FQMIKDELLR HDSGDQDINV VSTYISKMKV IYPAVEGRIK | |
Greater than 80% as determined by SDS-PAGE. |
4907 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |