missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L19, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L19, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6381-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055578.2 | |
39S ribosomal protein L19, mitochondrial Protein | |
Research Use Only | |
MRPL19 | |
Recombinant Protein | |
E. coli | |
MAACIAAGHW AAMGLGRSFQ AARTLLPPPA SIACRVHAGP VRQQSTGPSE PGAFQPPPKP VIVDKHRPVE PERRFLSPEF IPRRGRTDPL KFQIERKDML ERRKVLHIPE FYVGSILRVT TADPYASGKI SQFLGICIQR SGRGLGATFI LRNVIEGQGV EICFELYNPR VQEIQVVKLE KRLDDSLLYL RDALPEYSTF DVNMKPVVQE PNQKVPVNEL KVKMKPKPWS KRWERPNFNI KGIRFDLCLT EQQMKEAQKW NQPWLEFDMM REYDTSKIEA AIWKEIEASK RS | |
Greater than 90% as determined by SDS-PAGE. |
9801 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |