missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 14-3-3 eta / YWHAH Protein
A DNA sequence encoding the Homo sapiens (Human) 14-3-3 eta / YWHAH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP870-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003396.1 | |
14-3-3 eta / YWHAH Protein | |
Research Use Only | |
YWHAH | |
Recombinant Protein | |
E. coli | |
REQLLQRARL AEQAERYDDM ASAMKAVTEL NEPLSNEDRN LLSVAYKNVV GARRSSWRVI SSIEQKTMAD GNEKKLEKVK AYREKIEKEL ETVCNDVLSL LDKFLIKNCN DFQYESKVFY LKMKGDYYRY LAEVASGEKK NSVVEASEAA YKEAFEISKE QMQPTHPIRL GLALNFSVFY YEIQNAPEQA CLLAKQAFDD AIAELDTLNE DSYKDSTLIM QLLRDNLTLW TSDQQDEEAG EGN | |
Greater than 90% as determined by SDS-PAGE. |
7533 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |