missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 14-3-3 eta / YWHAH Protein

A DNA sequence encoding the Homo sapiens (Human) 14-3-3 eta / YWHAH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP870-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006021

  • 2235.00 EUR / 1mg

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003396.1
14-3-3 eta / YWHAH Protein
Research Use Only
YWHAH
Recombinant Protein
E. coli
REQLLQRARL AEQAERYDDM ASAMKAVTEL NEPLSNEDRN LLSVAYKNVV GARRSSWRVI SSIEQKTMAD GNEKKLEKVK AYREKIEKEL ETVCNDVLSL LDKFLIKNCN DFQYESKVFY LKMKGDYYRY LAEVASGEKK NSVVEASEAA YKEAFEISKE QMQPTHPIRL GLALNFSVFY YEIQNAPEQA CLLAKQAFDD AIAELDTLNE DSYKDSTLIM QLLRDNLTLW TSDQQDEEAG EGN
Greater than 90% as determined by SDS-PAGE.
7533
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers