missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Serum amyloid A Protein
A DNA sequence encoding the Equus caballus (Horse) Serum amyloid A protein, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Name | Serum amyloid A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | SAA1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024800
|
enQuireBio™
QP6647-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064800
|
enQuireBio™
QP6647-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034800
|
enQuireBio™
QP6647-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054800
|
enQuireBio™
QP6647-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074800
|
enQuireBio™
QP6647-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044800
|
enQuireBio™
QP6647-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Serum amyloid A Protein | |
Not determined. | |
Full Length | |
Horse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
Yeast | |
LLSFLGEAAR GTWDMIRAYN DMREANYIGA DKYFHARGNY DAAKRGPGGA WAAKVISDAR ENFQRFTDRF SFGGSGRGAE DSRADQAANE WGRSGKDPNH FRPHGLPDKY | |
Greater than 80% as determined by SDS-PAGE. |