missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-3 Protein

A cDNA sequence encoding the HCV NS5 Genotype-3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12192-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959648

  • 208.00 EUR / 100µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

Hepatitis C HCV NS5 Genotype-3 Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl, 1mM EDTA, 1.5M urea and 50% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
GST
HCV NS5 Genotype-3 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers