missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant H. pylori Vacuolating cytotoxin autotransporter Protein

A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Vacuolating cytotoxin autotransporter, was expressed in the hosts and tags indicated. option, abo

Brand:  enQuireBio™ QP8796-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066521

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_207680.1
Vacuolating cytotoxin autotransporter Protein
Research Use Only
vacA
Recombinant Protein
E. coli
TTVIIPAIVG GIATGAAVGT VSGLLGWGLK QAEEANKTPD KPDKVWRIQA GKGFNEFPNK EYDLYRSLLS SKIDGGWDWG NAATHYWVKG GQWNKLEVDM KDAVGTYNLS GLRNFTGGDL DVNMQKATLR LGQFNGNSFT SYKDSADRTT RVDFNAKNIL IDNFLEINNR VGSGAGRKAS STVLTLQASE GITSSKNAEI SLYDGATLN
Greater than 90% as determined by SDS-PAGE.
899415
500 μg
Not determined.
Partial (see sequence information for more details).
H. pylori
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers