missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori cagA Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) cagA, was expressed in the hosts and tags indicated. option, abo
Brand: enQuireBio™ QP8795-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_207343.1 | |
cagA Protein | |
Research Use Only | |
cagA | |
Recombinant Protein | |
Yeast | |
MTNETIDQTR TPDQTQSQTA FDPQQFINNL QVAFIKVDNV VASFDPDQKP IVDKNDRDNR QAFDGISQLR EEYSNKAIKN PTKKNQYFSD FIDKSNDLIN KDNLIDVESS TKSFQKFGDQ RYQIFTSWVS HQKDPSKINT RSIRNFMENI IQPPIPDDKE KAEFLKSAKQ SFAGIIIGNQ IRTDQKFMGV FDESLKERQE AEKNGGPTGG DWLDIFLSFI FNKKQSSDVK EAINQEPVPH VQPDIATTTT DIQGLPPEAR DLLDERGNFS KFTLGDMEML DVEGVADIDP NYKF | |
Greater than 90% as determined by SDS-PAGE. |
898777 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
H. pylori | |
His | |
Tris-based buffer, 50% glycerol |