missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Trans-activator protein BZLF1 Protein

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Trans-activator protein BZLF1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7277-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047970

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

YP_401673.1
Trans-activator protein BZLF1 Protein
Research Use Only
BZLF1
Recombinant Protein
E. coli
MMDPNSTSED VKFTPDPYQV PFVQAFDQAT RVYQDLGGPS QAPLPCVLWP VLPEPLPQGQ LTAYHVSTAP TGSWFSAPQP APENAYQAYA APQLFPVSDI TQNQQTNQAG GEAPQPGDNS TVQTAAAVVF ACPGANQGQQ LADIGVPQPA PVAAPARRTR KPQQPESLEE CDSELEIKRY KNRVASRKCR AKFKQLLQHY REVAAAKSSE NDRLRLLLKQ MCPSLDVDSI IPRTPDVLHE DLLNF
Greater than 90% as determined by SDS-PAGE.
3783744
50 μg
Not determined.
Full Length
Epstein-Barr
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers