missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein 1
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein H, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | YP_401700.1 |
---|---|
Gene ID (Entrez) | 3783742 |
Name | Envelope glycoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008740
|
enQuireBio™
QP7411-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028740
|
enQuireBio™
QP7411-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018740
|
enQuireBio™
QP7411-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038740
|
enQuireBio™
QP7411-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048740
|
enQuireBio™
QP7411-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058740
|
enQuireBio™
QP7411-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
YP_401700.1 | |
Envelope glycoprotein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3783742 | |
Research Use Only | |
gH | |
Recombinant Protein | |
E. coli | |
SLSEVKLHLD IEGHASHYTI PWTELMAKVP GLSPEALWRE ANVTEDLASM LNRYKLIYKT SGTLGIALAE PVDIPAVSEG SMQVDASKVH PGVISGLNSP ACMLSAPLEK QLFYYIGTML PNTRPHSYVF YQLRCHLSYV ALSINGDKFQ YTGAMTSKFL MGTYKRVTEK GDEHVLSLVF GKTKDLPDLR GPFSYPSLTS | |
Greater than 90% as determined by SDS-PAGE. |