missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein 1
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7278-ec-10ug
Additional Details : Weight : 0.01000kg
Specifica
YP_401678.1 | |
Envelope glycoprotein 1 | |
Research Use Only | |
gL | |
Recombinant Protein | |
E. coli | |
NWAYPCCHVT QLRAQHLLAL ENISDIYLVS NQTCDGFSLA SLNSPKNGSN QLVISRCANG LNVVSFFISI LKRSSSALTG HLRELLTTLE TLYGSFSVED LFGANLNRYA WHRGG | |
Greater than 90% as determined by SDS-PAGE. |
3783710 | |
10 μg | |
Not determined. | |
Full Length | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |