Learn More
enQuireBio™ Recombinant E. coli Transcriptional regulatory protein PhoP Protein
A DNA sequence encoding the Escherichia coli (strain K12) Transcriptional regulatory protein PhoP, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_415648.1 |
---|---|
Gene ID (Entrez) | 945697 |
Name | Transcriptional regulatory protein PhoP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088471
|
enQuireBio™
QP9558-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028481
|
enQuireBio™
QP9558-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098471
|
enQuireBio™
QP9558-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018481
|
enQuireBio™
QP9558-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038481
|
enQuireBio™
QP9558-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008481
|
enQuireBio™
QP9558-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_415648.1 | |
Transcriptional regulatory protein PhoP Protein | |
Not determined. | |
Full. Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945697 | |
Research Use Only | |
phoP | |
Recombinant Protein | |
Yeast | |
MRVLVVEDNA LLRHHLKVQI QDAGHQVDDA EDAKEADYYL NEHIPDIAIV DLGLPDEDGL SLIRRWRSND VSLPILVLTA RESWQDKVEV LSAGADDYVT KPFHIEEVMA RMQALMRRNS GLASQVISLP PFQVDLSRRE LSINDEVIKL TAFEYTIMET LIRNNGKVVS KDSLMLQLYP DAELRESHTI DVLMGRLRKK IQAQYPQEVI TTVRGQGYLF ELR | |
Greater than 80% as determined by SDS-PAGE. |