missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Transcription termination factor Rho Protein
A DNA sequence encoding the Escherichia coli (strain K12) Transcription termination factor Rho, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8868-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418230.1 | |
Transcription termination factor Rho Protein | |
Research Use Only | |
rho | |
Recombinant Protein | |
E. coli | |
MNLTELKNTP VSELITLGEN MGLENLARMR KQDIIFAILK QHAKSGEDIF GDGVLEILQD GFGFLRSADS SYLAGPDDIY VSPSQIRRFN LRTGDTISGK IRPPKEGERY FALLKVNEVN FDKPENARNK ILFENLTPLH ANSRLRMERG NGSTEDLTAR VLDLASPIGR GQRGLIVAPP KAGKTMLLQN IAQSIAYNHP DCVLMVLLID ERPEEVTEMQ RLVKGEVVAS TFDEPASRHV QVAEMVIEKA KRLVEHKKDV IILLDSITRL ARAYNTVVPA SGKVLTGGVD ANALHRPKRF FGAARNVEEG GSLTIIATAL IDTGSKMDEV IYEEFKGTGN MELHLSRKIA EKRVFPAIDY NRSGTRKEEL LTTQEELQKM WILRKIIHPM GEIDAMEFLI NKLAMTKTND DFFEMMKRS | |
Greater than 90% as determined by SDS-PAGE. |
948297 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |