missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Transcription termination factor Rho Protein

A DNA sequence encoding the Escherichia coli (strain K12) Transcription termination factor Rho, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8868-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046911

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_418230.1
Transcription termination factor Rho Protein
Research Use Only
rho
Recombinant Protein
E. coli
MNLTELKNTP VSELITLGEN MGLENLARMR KQDIIFAILK QHAKSGEDIF GDGVLEILQD GFGFLRSADS SYLAGPDDIY VSPSQIRRFN LRTGDTISGK IRPPKEGERY FALLKVNEVN FDKPENARNK ILFENLTPLH ANSRLRMERG NGSTEDLTAR VLDLASPIGR GQRGLIVAPP KAGKTMLLQN IAQSIAYNHP DCVLMVLLID ERPEEVTEMQ RLVKGEVVAS TFDEPASRHV QVAEMVIEKA KRLVEHKKDV IILLDSITRL ARAYNTVVPA SGKVLTGGVD ANALHRPKRF FGAARNVEEG GSLTIIATAL IDTGSKMDEV IYEEFKGTGN MELHLSRKIA EKRVFPAIDY NRSGTRKEEL LTTQEELQKM WILRKIIHPM GEIDAMEFLI NKLAMTKTND DFFEMMKRS
Greater than 90% as determined by SDS-PAGE.
948297
50 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers