missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Thioredoxin-1 Protein

A DNA sequence encoding the Escherichia coli (strain K12) Thioredoxin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8944-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007271

  • 309.00 EUR / 10µg


Estimated Shipment
31-07-2020
Add to basket

Specifications

Specifications

NP_418228.2
Thioredoxin-1 Protein
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
31656514
Greater than 90% as determined by SDS-PAGE.
Research Use Only
trxA
Recombinant Protein
E. coli
SDKIIHLTDD SFDTDVLKAD GAILVDFWAE WCGPCKMIAP ILDEIADEYQ GKLTVAKLNI DQNPGTAPKY GIRGIPTLLL FKNGEVAATK VGALSKGQLK EFLDANLA
Videos
Documents