missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli tetR Protein
A DNA sequence encoding the Escherichia coli tetR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7416-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000088605.1 | |
tetR Protein | |
Research Use Only | |
tetR | |
Recombinant Protein | |
E. coli | |
MSRLDKSKVI NSALELLNEV GIEGLTTRKL AQKLGVEQPT LYWHVKNKRA LLDALAIEML DRHHTHFCPL EGESWQDFLR NNAKSFRCAL LSHRDGAKVH LGTRPTEKQY ETLENQLAFL CQQGFSLENA LYALSAVGHF TLGCVLEDQE HQVAKEERET PTTDSMPPLL RQAIELFDHQ GAEPAFLFGL ELIICGLEKQ LKCESGS | |
Greater than 90% as determined by SDS-PAGE. |
31515962 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |