missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli tetR Protein

A DNA sequence encoding the Escherichia coli tetR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7416-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028770

  • 2985.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_000088605.1
tetR Protein
Research Use Only
tetR
Recombinant Protein
E. coli
MSRLDKSKVI NSALELLNEV GIEGLTTRKL AQKLGVEQPT LYWHVKNKRA LLDALAIEML DRHHTHFCPL EGESWQDFLR NNAKSFRCAL LSHRDGAKVH LGTRPTEKQY ETLENQLAFL CQQGFSLENA LYALSAVGHF TLGCVLEDQE HQVAKEERET PTTDSMPPLL RQAIELFDHQ GAEPAFLFGL ELIICGLEKQ LKCESGS
Greater than 90% as determined by SDS-PAGE.
31515962
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions