missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli ssbF Protein

A DNA sequence encoding the Escherichia coli (strain K12) ssbF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7083-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027330

  • 1550.00 EUR / 200µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_061439.1
ssbF Protein
Research Use Only
ssbF
Recombinant Protein
E. coli
AVRGINKVIL VGRLGKDPEV RYIPNGGAVA NLQVATSESW RDKQTGEMRE QTEWHRVVLF GKLAEVAGEC LRKGAQVYIE GQLRTRSWED NGITRYVTEI LVKTTGTMQM LVRAAGAQTQ PEEGQQFSGQ PQPEPQAEAG TKKGGAKTKG RGRKAAQPEP QPQPPEGDDY GFSDDIPF
Greater than 90% as determined by SDS-PAGE.
1263584
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers