missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli plsC Protein

A DNA sequence encoding the Escherichia coli (strain K12) plsC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7206-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077700

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_417490.1
plsC Protein
Research Use Only
plsC
Recombinant Protein
Yeast
MLYIFRLIIT VIYSILVCVF GSIYCLFSPR NPKHVATFGH MFGRLAPLFG LKVECRKPTD AESYGNAIYI ANHQNNYDMV TASNIVQPPT VTVGKKSLLW IPFFGQLYWL TGNLLIDRNN RTKAHGTIAE VVNHFKKRRI SIWMFPEGTR SRGRGLLPFK TGAFHAAIAA GVPIIPVCVS TTSNKINLNR LHNGLVIVEM LPPIDVSQYG KDQVRELAAH CRSIMEQKIA ELDKEVAERE AAGKV
Greater than 90% as determined by SDS-PAGE.
947496
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers