missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Phosphoenolpyruvate carboxylase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Phosphoenolpyruvate carboxylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8934-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418391.1 | |
Phosphoenolpyruvate carboxylase Protein | |
Research Use Only | |
ppc | |
Recombinant Protein | |
E. coli | |
YSALRSNVSM LGKVLGETIK DALGEHILER VETIRKLSKS SRAGNDANRQ ELLTTLQNLS NDELLPVARA FSQFLNLANT AEQYHSISPK GEAASNPEVI ARTLRKLKNQ PELSEDTIKK AVESLSLELV LTAHPTEITR RTLIHKMVEV NACLKQLDNK DIADYEHNQL MRRLRQLIAQ SWHTDEIRKL RPSPVDEAKW GFAVVENSLW QGVPNYLREL NEQLEENLGY KLPVEFVPVR FTSWMGGDRD GNPNVTADIT RHVLLLSRWK ATDLFLKDIQ VLVSELSMVE ATPELLALVG EEGAAEPYRY LMKNLRSRLM ATQAWLEARL KGEELPKPEG LLTQNEELWE PLYACYQSLQ ACGMGIIANG DLLDTLRRVK CFGVPLVRID IRQESTRHTE ALGELTRYLG IGDYESWSEA DKQAFLIREL NSKRPLLPRN WQPSAETREV LDTCQVIAEA PQGSIAAYVI SMAKTPSDVL AVHLLLKEAG IGFAMPVAPL FETLDDLNNA NDVMTQLLNI DWYRGLIQGK QMVMIGYSDS AKDAGVMAAS WAQYQAQDAL IKTCEKAGIE LTLFHGRGGS IGRGGAPAHA ALLSQPPGSL KGGLRVTEQG EMIRFKYGLP EITVSSLSLY TGAILEANLL PPPEPKESWR RIMDELSVIS CDVYRGYVRE NKDFVPYFRS ATPEQELGKL PLGSRPAKRR PTGGVESLRA IPWIFAWTQN RLMLPAWLGA GTALQKVVED GKQSELEAMC RDWPFFSTRL GMLEMVFAKA DLWLAEYYDQ RLVDKALWPL GKELRNLQEE DIKVVLAIAN DSHLMADLPW IAESIQLRNI YTDPLNVLQA ELLHRSRQAE KEGQEPDPRV EQALMVTIAG IAAGMRNTG | |
Greater than 90% as determined by SDS-PAGE. |
948457 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |